Learn More
Spectrum™ Dialysis Reservoir
Provides the optimum environment for laboratory dialysis
Manufacturer:  Spectrum™ 132005
 View more versions of this product
Catalog No. 08-750-20B
Add to cart
Includes: Silent Swirl stir bar
Description
Description
Stir bar generates vortex to draw dialysis tubing into the center, improving efficiency.
  • 600mL: For RDDs up to 1mL and tubing flat widths up to 16mm
  • 1800mL: For any size RDD and tubing flat widths up to 80mm
For use with Ready-to-Use Dialysis Device (RDD) or standard dialysis tubing
Specifications
Specifications
1800mL
115mm
11.8 in.
Polystyrene
Each
4.5 in.
Provide the optimum environment for laboratory dialysis
300mm
Dialysis Reservoir
Silent swirl stir bar