Learn More
Spectrum™ Spectrum™ Dialysis Tubing Closures: Spectra/Por™ Standard Type
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
$229.50 - $329.00
Catalog Number | Mfr. No. | Color | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Width (Metric) | Price | Quantity & Availability | ||||
08-700-265
|
Spectrum™
142937 |
Blue | 55 mm |
Pack of 10 for $329.00
|
|
||||
08-801-36
|
Spectrum™
142936 |
Blue | 35 mm |
Pack of 10 for $316.00
|
|
||||
08-700-254
|
Spectrum™
142738 |
White | 75 mm |
Pack of 10 for $329.00
|
|
||||
08-700-260
|
Spectrum™
142838 |
Green | 75 mm |
Pack of 10 for $329.00
|
|
||||
08-700-253
|
Spectrum™
142737 |
White | 55 mm |
Pack of 10 for $329.00
|
|
||||
08-700-267
|
Spectrum™
142939 |
Blue | 90 mm |
Pack of 10 for $229.50
|
|
||||
08-700-255
|
Spectrum™
142739 |
White | 90 mm |
Pack of 10 for $229.50
|
|
||||
08-700-266
|
Spectrum™
142938 |
Blue | 75 mm |
Pack of 10 for $329.00
|
|
||||
08-670-11AA
|
Spectrum™
132734 |
Orange | 12 mm |
Pack of 10 for $267.00
|
|
||||
08-700-261
|
Spectrum™
142839 |
Green | 90 mm |
Pack of 10 for $229.50
|
|
||||
08-801-35
|
Spectrum™
142736 |
White | 35 mm |
Pack of 10 for $312.00
|
|
||||
08-750-24B
|
Spectrum™
132735 |
Orange | 23 mm |
Pack of 10 for $290.50
|
|
||||
08-670-11B
|
Spectrum™
132738 |
Orange | 75 mm |
Pack of 10 for $329.00
|
|
||||
08-801-11
|
Spectrum™
142836 |
Green | 35 mm |
Pack of 10 for $299.00
|
|
||||
08-750-24C
|
Spectrum™
132737 |
Orange | 55 mm |
Pack of 10 for $329.00
|
|
||||
08-671-87
|
Spectrum™
142837 |
Green | 55 mm |
Pack of 10 for $309.00
|
|
||||
08-670-11A
|
Spectrum™
132736 |
Orange | 35 mm |
Pack of 10 for $329.00
|
|
||||
Description
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Spectra/Por Standard Closure Type:
- Made from polypropylene to aid in sample buoyancy
- Maintains sample vertical orientation when used with a Spectra/Por Weighted Closure.
- 5 sealing width sizes: 12mm, 23mm, 35mm, 55mm and 75mm
- 4 colors available for easier sample identification
- Autoclavable
- 10/pkg; Use with RC dialysis tubing only. Not for use with CE tubing.
Specifications
Autoclavable |