Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Drawer/Ganizers™ Drawer Organizer Units

Handy compartmentalized trays keep each item in its proper place

Manufacturer:  Spectrum™ 144465

 View more versions of this product

Catalog No. 14-334-13A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • No need to rummage through cluttered, messy drawers
  • Constructed of heavy-duty polystyrene
  • Fit drawers of all standard modular metal lab furniture
  • Withstand heavy use and abuse

Specifications

Specifications

17.36 x 19.37 x 2.87 in. (44.1 x 49.2 x 7.3cm)
For bulky items that won't fit other drawer spacers, e.g., mortars and pestles, watch Glasses, filter Paper, coils of tubing, small appliances and instruments, centrifuge heads and rotors
7.3cm
44.1cm
Utility Spacer
4.0
49.2cm
Refrigerators and bench tops
2.87 in.
17.36 in.
PS (Polystyrene)
L
19.37 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit