Spectrum™ DynaGard™ Syringe Filter Tips
With looped hollow fiber membrane to maximize surface area and reduce fouling
Manufacturer: Spectrum™ DG2M330100
 View more versions of this product
Catalog No. 08-671-8F
  Check Availability
Add to Cart
    Specifications
    Inlet Style Female Luer Lok
    Pore Size 0.2μm
    Volume (Metric) 5 to 20mL
    Outlet Style Male Luer Taper
    Sterile No
    Type Syringe Filter Tips
    Material Mixed Cellulose Esters (MCE)
  • Description & Specifications
Related Products
Product Content Feedback