Learn More
Spectrum™ DynaGard™ Syringe Filter Tips
With looped hollow fiber membrane to maximize surface area and reduce fouling
Manufacturer:  Spectrum™ DG2P110200
 View more versions of this product
Catalog No. 08-671-6B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Narrow housing design minimizes hold-up volume and enables easy aspiration and dispensing in ampules, test tubes and other small vessels
  • Available with either 0.2µm hydrophilic mixed cellulose ester (ME) fibers for use with aqueous solutions or 0.2µm hydrophobic polypropylene (PP) fibers for use with organic solvents and alcohols
Specifications
Specifications
Non-sterile
Polypropylene (PP)
Female Luer Lok
0.2μm
Syringe Filter Tips
<5mL
Male Luer Taper