Learn More
Spectrum™ Spectrum™ Dialysis Tubing Closures: Spectra/Por™ Standard Type
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Manufacturer:  Spectrum™ 132735
 View more versions of this product
Catalog No. 08-750-24B
Add to cart
Description
Description
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Spectra/Por Standard Closure Type:
  • Made from polypropylene to aid in sample buoyancy
  • Maintains sample vertical orientation when used with a Spectra/Por Weighted Closure.
  • 5 sealing width sizes: 12mm, 23mm, 35mm, 55mm and 75mm
  • 4 colors available for easier sample identification
  • Autoclavable
  • 10/pkg; Use with RC dialysis tubing only. Not for use with CE tubing.
Specifications
Specifications
Yes
For sample buoyancy and vertical orientation, Spectrum laboratories, use with RC only
32° to 250°F
Regenerated Cellulose
Each
Orange
Polypropylene
0° to 121°C
Dialysis Tubing Closure