Learn More
Spectrum™ Spectrum™ Drawer/Ganizer™ Portable Organizer Trays
Rigid 1/8 in. polystyrene trays with easy-gripping folded edges for transporting lab items and mobilizing experimental activities
Manufacturer:  Spectrum™ 155450
 View more versions of this product
Catalog No. 08-607-055
Add to cart
Description
Description
Rigid 1/8 in. polystyrene trays with easy-gripping folded edges for transporting lab items and mobilizing experimental activities
Deep compartments are ideal for holding large quantities or providing secondary containment for spills. Also designed to fit lab drawers, refrigerators and bench tops to protect supplies, organize valuable space and promote laboratory safety.
  • Designed for portability and storage in drawers
  • Protects supplies, promotes safety and provides secondary containment
  • Six tray configurations available in two sizes (large and small)
Specifications
Specifications
15 Various size compartments
0.32cm
Parts and Chromatography Tray
Large
Polystyrene