Learn More
Spectrum™ Spectra Mesh™ Woven, Nylon Filters
Manufacturer:  Spectrum™ 145809
 View more versions of this product
Catalog No. 08-671-90
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
64μm
Circle
30μm
Nylon
55mm
Hazardous waste, drinking water purification, Acid rain measurement, suspended soils, air/asbestos, coal dust analysis, oceanography, trace element analysis