Learn More
Spectrum™ Spectrum™ Spectra Mesh™ Woven, Polyester Filters
Consist of individual strands woven into a mesh screen
Manufacturer:  Spectrum™ 148242
 View more versions of this product
Catalog No. 08-801-23
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Polyester
25mm
Drinking water purification, suspended soils, air/asbestos, coal dust analysis, trace element analysis, fertilizers, aqueous serum clarification
Circle
10μm
55μm