Learn More
Spectrum™ Spectra Mesh™ Woven, Polyester Filters
Manufacturer:  Spectrum™ 145831
 View more versions of this product
Catalog No. 08-671-91
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
55μm
Circle
10μm
Polyester
55mm
Drinking water purification, suspended soils, air/asbestos, coal dust analysis, trace element analysis, fertilizers, aqueous serum clarification, organic serum clarification