Learn More
Spectrum™ Spectra/Por™ 1-4 Standard RC Dialysis Membrane in Flat Sheets
Clear, flexible and sturdy Standard RC (regenerated cellulose) in single layer square sheets that can be used with a variety of dialysis equipment or can be cut into specific dimensions for attaching to support frames for customized diagnostic assays, equilibrium studies, desalting, protein purification, etc.
Manufacturer:  Spectrum™ 132686
 View more versions of this product
Catalog No. 08-670-4
Add to cart
Description
Description
  • Flat-Sheets deal for use with dialysis equipment or cutting to size
  • Available in 3.5 kD, 6-8 kD and 12-14 kD MWCO's
  • Good chemical compatibility with dilute strong acids & bases, conc. weak acids and bases, many alcohols & some organics.
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • 25 sheets/pkg
Do not tie knots in tubing; use closures
Specifications
Specifications
Clear
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
200mm
39° to 250°F
7.8 in.
Spectra/Por™ 2
25 Sheets
7.8 x 7.8 in. (200 x 200mm)
7.8 in.
12 to 14 kd
4° to 121°C
200mm
Spectra/Por™ 2 Dialysis Membrane Flat Sheets