Learn More
Spectrum™ Labs Spectra/Por™ 2 and 4 12-14 kD MWCO Standard RC Dry Dialysis Kits
For smaller dialysis experiments or evaluating dialysis tubing for dialysis application
Manufacturer:  Spectrum™ 132684T1
 View more versions of this product
Catalog No. 08-700-157
Add to cart
Includes:
  • 1m or 5m of Spectra/Por 2 or 4 dialysis tubing
  • One Standard Spectra/Por Closure (for buoyance)
  • One weighted Spectra/Por Closure (for orientation)
  • Two adhesive tubing openers
Description
Description
  • Hydrophilic, symmetric porosity
  • Ensures higher permeability and basic dialysis
  • Available as dry with glycerin (soak in water)
Specifications
Specifications
46mL/cm vol./Length
76mm
1m tubing/kit
12-14 kD