Learn More
Spectrum™ Labs Spectra/Por™ 7 15,000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
Manufacturer:  Spectrum™ 132124T
 View more versions of this product
Catalog No. 08-700-218
Add to cart
Includes:
  • 1meter of pre-treated tubing, a pair of closures and 5 opening picks
Description
Description
  • Hydrophilic, symmetric porositiy
  • Equilibrated in water
  • Packaged wet in preservative solution of 0.05% sodium azide
  • For small experiments or qualifying a membrane for a particular application
Specifications
Specifications
6.4mL/cm vol./Length
29mm
1m tubing/kit
15 kD