Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrum™ MediaKap™ and MediaKap Plus HF Filters
0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling
Supplier: Spectrum™ MP2M50B03S
Description
- Efficiently sterilize and clarify culture medium or buffer solution
- Smaller, narrow design for low hold-up volume
- Polypropylene vent prevents air lock
- Operates with peristaltic pump, vacuum, or syringe
- Available in five different sizes that filter designated volume in 15 to 20 minutes
- Sterile, biocompatible and nonpyrogenic
- USP Class VI; 100% integrity tested
- Optional filling bell maintains sterility while filling bottles
- MediaKap Plus filters utilize an advanced DynaFibre membrane that further increases filtration efficiency and dramatically reduces the time required to filter serum-enriched medium
Specifications
VarHB/VarHB | |
Polycarbonate, Polyurethane | |
0.2 μm | |
440 cm2 | |
Efficiently sterilize and clarify culture medium or buffer solution | |
3 Case | |
25 to 50 L |
MediaKap-50 Plus | |
MCE | |
Sterile | |
2000 mL/min. at 10psig | |
Automatic vent to prevent air lock, filling bell | |
Bottle Top Filter |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction