Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ MediaKap™ and MediaKap Plus HF Filters

0.2µm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling

Manufacturer:  Spectrum™ MP2M25B06S

 View more versions of this product

Catalog No. 08-671-4F


  Check Availability
Add to cart

Specifications

Description

  • Efficiently sterilize and clarify culture medium or buffer solution
  • Smaller, narrow design for low hold-up volume
  • Polypropylene vent prevents air lock
  • Operates with peristaltic pump, vacuum, or syringe
  • Available in five different sizes that filter designated volume in 15 to 20 minutes
  • Sterile, biocompatible and nonpyrogenic
  • USP Class VI; 100% integrity tested

  • Optional filling bell maintains sterility while filling bottles
  • MediaKap Plus filters utilize an advanced DynaFibre membrane that further increases filtration efficiency and dramatically reduces the time required to filter serum-enriched medium

Description & Specifications

Specifications

Yes
6 Case
VarHB/VarHB
MediaKap-25 Plus
1400mL/min. at 10psi
Automatic vent to prevent air lock, filling bell
10 to 25L
0.2μm
Polycarbonate, Polyurethane
Bottle Top Filter Unit
185cm2
Efficiently sterilize and clarify culture medium or buffer solution
Mixed Cellulose Ester
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit