Learn More
Spectrum™ Spectra/Mesh Woven Polymer Macrofiltration Squares: N (Nylon)
Combines high tensile strength with flexibility
Manufacturer:  Spectrum™ 146506
 View more versions of this product
Catalog No. 08-670-203
Add to cart
Description
Description
Combines high tensile strength with flexibility
Ideal for straining, filtering and screening solutions or suspensions containing high-molecular-weight molecules or particles.
  • Nonabsorbent
  • Combines high tensile strength with flexibility
Specifications
Specifications
4.65 in2
Sterile
64μm
3 to 10
ISO 9001:2008
21%
30μm
180°C
Spectra Mesh™ Woven Filter
High
Hydrophilic
Nylon