Learn More
Spectrum™ Spectra/Por™ 1 RC Dialysis Membrane Tubing 6000 to 8000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Manufacturer:  Spectrum™ 132650
 View more versions of this product
Catalog No. 08-670A
Add to cart
Description
Description
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Specifications
Specifications
0.57 in.
100 ft.
Spectra/Por™ Dialysis Tubing
4° to 121°C
Regenerated Cellulose
Each
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
30.5m
39.2° to 249.8°F
14.6mm
Contain glycerin as a humectant
6000 to 8000 d