Learn More
Spectrum™ Spectra/Por™ 2 RC Dialysis Membrane Tubing 12,000 to 14,000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Manufacturer:  Spectrum™ 132680
 View more versions of this product
Catalog No. 08-670-3A
Add to cart
Description
Description
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Specifications
Specifications
1.14 in.
49.2 ft.
Spectra/Por™ 2 Dialysis Tubing
4° to 121°C
Regenerated Cellulose
Each
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
15m
39.2° to 249.8°F
29mm
Contain glycerin as a humectant
12 to 14 kd