Learn More
Spectrum™ Spectra/Por™ 7 Membrane Tubing (Sulfur and Heavy Metal Free):1 1000 to 50,000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Manufacturer:  Spectrum™ 132116
 View more versions of this product
Catalog No. 08-680-5G
Add to cart
This item is not returnable. View return policy
Description
Description
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Specifications
Specifications
Transparent
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
5m
39.2° to 249.8°F
25.5mm
Packaged wet in 0.05% sodium azide
8000 d
1 in.
16.4 ft.
Spectra/Por™ 7 Membrane Tubing
4° to 121°C
Regenerated Cellulose
Each