Learn More
Spectrum™ Spectra/Por™ Weighted Closures
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Manufacturer:  Spectrum™ 132743
 View more versions of this product
Catalog No. 08-750-24F
Add to cart
Description
Description
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Spectra/Por Weighted Closure Type:
  • Made from polypropylene with a fluorocarbon coated SS bar to anchor tubing downward
  • Use with buoyant Standard Closure to maintain vertical sample orientation.
  • 5 sealing width sizes: 12mm, 23mm, 35mm, 55mm and 75mm
  • Autoclavable
  • 10/pkg; Use with RC dialysis tubing only. Not for use with CE tubing.
Specifications
Specifications
32° to 250°F
Regenerated Cellulose
Yes
White
Use with buoyant standard closure to maintain vertical sample orientation, use with RC dialysis tubing only, not for use with CE tubing
10 Pack
0° to 121°C
Dialysis Tubing Closure
Weighted
23mm
Polypropylene
Fluorocarbon coated stainless steel bar to anchor tubing downward