Learn More
Spectrum™ Spectra/Por™ Weighted Closures
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Manufacturer:  Spectrum™ 132746
 View more versions of this product
Catalog No. 08-750-24J
Add to cart
Description
Description
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Specifications
Specifications
Yes
White
Use with buoyant standard closure to maintain vertical sample orientation, use with RC dialysis tubing only, not for use with CE tubing
32° to 250°F
Regenerated Cellulose
10 Pack
Weighted
75mm
Polypropylene
0° to 121°C
Dialysis Tubing Closure
Fluorocarbon coated stainless steel bar to anchor tubing downward