Learn More
Spectrum™ Spectra/Por™ Weighted Paired Closures
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Manufacturer:  Spectrum™ 132749
 View more versions of this product
Catalog No. 08-750-24L
Add to cart
Description
Description
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
  • 10 pairs (20 closures)/pkg
Specifications
Specifications
Yes
Orange or White
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
32° to 250°F
Regenerated Cellulose
20 Pack
Paired
12mm
Polypropylene
0° to 121°C
Dialysis Tubing Closure