Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Standard RC Dialysis Tubing

Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes

Manufacturer:  Spectrum™ 132706

 View more versions of this product

Catalog No. 08-670-247

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

  • Membrane grade
  • Membrane type: RC
  • Molecular weight cut-off: 12 to 14K

Description & Specifications

Specifications

1.14 in.
100 ft.
Spectra/Por™ 4 Dialysis Tubing
4° to 121°C
Regenerated Cellulose
100 Pack
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
30m
39.2° to 249.8°F
29mm
Contain glycerin as a humectant
12 to 14 kd
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit