Learn More
3M™ Speedglas™ Heavy-Duty Welding Helmet with Welding Filter G5-01VC V-100 Vortex™ Cooling Valve Assembly, ADF
Heavy-duty welding helmet filter variable color technology allows welders to select between 3 different color options (clear, cool, or warm), improving ability to see the weld puddle, joint, and arc.
Supplier: 3M™ 7100214379
Description
- Dark shade options include 5, 8-14
- Natural Color technology makes filter view seem lighter, more detailed and colorful
- Auto-ON/OFF
- Large (104 x 170 mm), clear protective visor is curved, with a variety of ideal views
- G5-01 helmet airflow optimization system lets users direct the airflow to either the face, visor, or somewhere in-between.
- Re-configure the helmet using additional accessories to find the right coverage and protection. Optional accessories include: task light assembly, 1000 Assigned Protection Factor (APF) assembly, high-visibility head cover, leather head/neck cover, and more.
- Supplied Air Respirator Vortex™ Cooling Assembly V-100 may cool the air up to 50°F (28°C)
- Slim profile and ergonomic design makes the helmet lightweight and easy to utilize in tight welding spaces
- Bluetooth enabled
- Utilizing the 3M Connected Equipment app, users get access to additional functionalities, statistics, and documentation
Specifications
ANSI Z87.1-2015; NIOSH Approved | |
SAR Welding System | |
G5 Series storage/carry bag | |
1 Ea. |
Black | |
Grinding, Welding | |
70071697240 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.