Learn More
3M™ Speedglas™ Inner Grinding Visor, Anti-Fog and Hardcoated
Grinding Visor comes with premium anti-fog and scratch resistant coatings
Manufacturer:  3M™ 70071735677
Catalog No. 19-072-134
Add to cart
Description
Description
  • Regularly clean with a soft cloth and replace if any pits or scratches occur.
  • For maximum safety and comfort, make it a habit to check equipment regularly.
  • Replace worn out or damaged parts.
Specifications
Specifications
Clear
Visors
Anti-Fog and Scratch Resistant
Metal Repair, Metal Processing, Welding, Grinding
5/Case