Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPEN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10058799
Description
SPEN Polyclonal specifically detects SPEN in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
SPEN | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
HIAA0929, KIAA0929nuclear receptor transcription cofactor, MINTMsx2 interacting nuclear target (MINT) homolog, RBM15C, SHARPmsx2-interacting protein, SMART/HDAC1 associated repressor protein, SMART/HDAC1-associated repressor protein, SPEN homolog, spen homolog, transcriptional regulator (Drosophila) | |
Rabbit | |
Affinity Purified | |
RUO | |
23013 | |
Human | |
IgG |
Western Blot, Immunoprecipitation, Proximity Ligation Assay, ChIP Assay | |
1.0 mg/mL | |
Western Blot Reported in scientific literature (PMID: 29880719)., Immunoprecipitation 2 - 5 ug/mg lysate, Proximity Ligation Assay Reported in scientific literature (PMID: 29880719)., Chromatin Immunoprecipitation (ChIP) 2-5 ug/mg lysate | |
NP_055816.2 | |
SPEN | |
The immunogen recognized by this antibody maps to a region between residue 3375 and 3425 of human SMART/HDAC1 associated repressor protein (spen homolog, transcriptional regulator) using the numbering given in entry NP_055816.2 (GeneID 23013). | |
100 μL | |
Primary | |
Expected to cross react based on sequence identity: Orangutan, Rhesus Monkey, Chimpanzee, Northern white-cheeked gibbon, Crab-eating macaque. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction