Learn More
Mini Environmental Quality Meters
SCI_ED
Monitor up to 12 environmental parameters with one tiny instrument
Specifications
Certifications/Compliance CE
Includes Wrist strap, carry pouch, battery and instructions
Test Time 5 sec.
View More Specs
Includes: Wrist strap, carry pouch, and button cell battery providing at least 40 hours of continuous use
Products 1
Catalog Number Mfr. No. Type For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Type For Use With (Application) Price Quantity & Availability  
S07073
SCI_ED
View Documents
Sper Scientific
850027
12-Paramaters Mini Environmental Quality Meter Monitors Wind Speed, Air Temperature, Air Volume, Relative Humidity, Wind Chill, Heat Stress Index, Dew Point, Wet Bulb, PT 1000 ohm Probe, Baromatric Pressure, Altitutde, Visible Light and Compass This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Lightweight, tiny size with simple, one-hand operation.
  • Senses airflow as low as 0.4 m/s with an accuracy of ±3% (4% >20m/s)
  • High precision humidity sensor with fast response time
  • Built-in microprocessor circuit assures excellent performance and accuracy
  • Concise and compact buttons arrangement for easy operation
  • Maximum and minimum memory with recall
  • Hold function
  • °C/°F selection
  • Wristlet design provides extra protection to the instrument
  • Low-friction ball bearing mounted impeller design provides high accuracy at high and low air velocity
  • Replaceable snap-in anemometer vane (not included)
  • For accessories, see cat. no. S07071
Specifications
Specifications
CE
Wrist strap, carry pouch, battery and instructions
5 sec.