Learn More
Honeywell™ Sperian™ Freedom Disposable Respirators
Ideal for occasional users
Manufacturer:  Honeywell™ 14150097
 View more versions of this product
Catalog No. 19-059-476
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Available in multiple models for a variety of applications
  • Three sizes ensure a secure, comfortable fit
  • Choose from contaminant-specific models or multi-contaminant, multi-use respirators (with cartridge and filter pre-assembled), designed to meet NIOSH standards for multiple contaminant exposure
  • Dispose when cartridges are expended, replacement parts and decontamination concerns are eliminated
Order Info
See chart below for a complete selection of respirators with pre-attached cartridges and filters. Probed models are also available; contact your Fisher Scientific™ Customer Service Representative for details. (Do not exceed maximum use concentration established by regulatory standards.)
Specifications
Specifications
Safety
NIOSH 42CFR84
P100 particulates
Thermoplastic elastomer
Thermoplastic elastomer
1
Single strap with cradle
0.34kg
Green
Yes
Fire protection, medical, pharmaceutical, transportation
Reusable Bag
Half Mask Respirator
Medium
0.75 lb.