Sperian One-Fit Healthcare N95 Particulate Respirator and Surgical Mask Get IT Item

Fit most face shapes and sizes while offering the ultimate in protection and comfort for healthcare settings

Manufacturer: Honeywell Safety Products 14110451

 View more versions of this product

Catalog No. 19-999-401

  Check Availability
Add to Cart

    Specifications

    Certifications NIOSH, CDC
    Quantity 20
    Disposable Yes
    Filter Type N95 Filter
    For Use With (Application) Designed for healthcare settings and use in emergency rooms, operating rooms and intensive care units
    Size Universal
    Product Type Flat form
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit