Learn More
Sperian SAF-T-FIT™ P95 Particulate Respirators
For use with contaminant aerosols containing oil
Manufacturer:  Honeywell Safety Products 14110430
 View more versions of this product
Catalog No. 19-034-917
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
10 Pack
Yes
Contaminant aerosols containing oil
Medium/Large
P1135-1
P95 Filter
Yes
NIOSH TC-84A-3879