Sperian SAF-T-FIT™ P95 Particulate Respirators

For use with contaminant aerosols containing oil

Manufacturer: Honeywell Safety Products 14110426

 View more versions of this product

Catalog No. 19-034-913

  Check Availability
Add to Cart

    Specifications

    Certifications NIOSH TC-84A-3877
    Quantity 20 Pack
    Disposable Yes
    Filter Type P95 Filter
    For Use With (Application) Contaminant aerosols containing oil
    Latex Free Yes
    Size Medium/Large
    Product Type P1115
    Color Code White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit