Learn More
Sperian™ SAF-T-FIT™ Plus N95 Particulate Respirators
For use with particulate aerosols free of oil
Manufacturer:  Honeywell Safety Products 14110391
 View more versions of this product
Catalog No. 19-034-903
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
20 Pack
95% against solid particulates and non-petroleum based liquid aerosols
White
Yes
Welding, soldering, brazing, torch cutting, metal pouring, grinding, sanding, particulate aerosols free of oil
Yes
Medium/Large
N1115
Molded cup
Green
N95 Filter
Boomerang nose seal and valve
Humidity
NIOSH TC-84A-3769