Sperian™ SAF-T-FIT™ Plus N95 Particulate Respirators

For use with particulate aerosols free of oil

Manufacturer: Honeywell Safety Products 14110396

 View more versions of this product

Catalog No. 19-034-905

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Certifications NIOSH TC-84A-3745
    Quantity 10 Pack
    Disposable Yes
    Filter Type N95 Filter
    For Use With (Application) Welding, soldering, brazing, torch cutting, metal pouring, grinding, sanding, particulate aerosols free of oil
    Includes Boomerang nose seal and valve
    Latex Free Yes
    Resistant To Humidity
    Size Small
    Product Type N1125Ov
    Filter Efficiency 95% against solid particulates and non-petroleum based liquid aerosols
    Design Type Molded cup
    Color Gray
    Color Code Red

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit