Learn More
Sperian™ SAF-T-FIT™ Plus N95 Particulate Respirators
For use with particulate aerosols free of oil
Manufacturer:  Honeywell Safety Products 14110398
 View more versions of this product
Catalog No. 19-034-907
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
10 Pack
95% against solid particulates and non-petroleum based liquid aerosols
Gray
Yes
Welding, soldering, brazing, torch cutting, metal pouring, grinding, sanding, particulate aerosols free of oil
Yes
XLarge
N1125Ov
Molded cup
Blue
N95 Filter
Boomerang nose seal and valve
Humidity
NIOSH TC-84A-3745