Sperian™ SAF-T-FIT™ Plus P100 Respirators

Suggested for use against oil and non-oil based particulates

Manufacturer: Honeywell Safety Products 14110440

 View more versions of this product

Catalog No. 19-166-846

  Check Availability
Add to Cart

    Specifications

    Certifications NIOSH
    Quantity Each
    Disposable Yes
    Filter Type P100 Filter
    For Use With (Application) Full sealing flange for superior fit and protection, against oil and non-oil based particulates
    Includes Boomerang nose seal, full face seal and valve
    Latex Free Yes
    Size Medium/Large
    Product Type P100 Respirator
    Color White
    Color Code Green

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit