Learn More
Honeywell™ Sperian™ Survivair 2000 Series Half-Mask Respirators
The 2000 series silicone reusable half-mask respirator features a lightweight and innovative yoke design with capability to convert to a CF-SAR.
Manufacturer:  Honeywell™ B260000
 View more versions of this product
Catalog No. 18-245B
Add to cart
Description
Description
  • Head harness designed to reduce facepiece slippage
  • Silicone facepiece construction is lightweight and durable
  • Rear split strap for easy donning and better support
  • Compatible with S-Series filters and cartridges
Specifications
Specifications
NIOSH 42CFR84
Additional sealing surface, improved self-adjusting nosebridge area and yoke-design headstrap
Half Mask Respirator
Blue
Silicone
Medium