Learn More
Honeywell™ Sperian™ S-Series Cartridges and Filters
S-Series filters work with both full and half mask respirators that only accept threaded cartridge connections
Manufacturer:  Honeywell™ 105310
 View more versions of this product
Catalog No. 18-265A
Add to cart
Description
Description
Survivair S-Series Screw-On Cartridges thread easy for tightening cartridge on seal.
Safety/5, † OSHA regulations require gasproof goggles be worn when half-mask respirators are used for protection against formaldehyde.
Specifications
Specifications
NIOSH
For protection against organic vapors
Cartridge Filter
Threaded
99.97%
4 Pack
Half mask and full facepiece respirator
Yellow, Magenta
Organic Vapor, Acid Gas Cartridge
P100 Filter