Sperian Survivair W4K Constant Flow Supplied Air Pump System Accessories

Manufacturer: Honeywell Safety Products 14900417

 View more versions of this product

Catalog No. 17-638-3

  Check Availability
Add to Cart

    Specifications

    Product Type Schrader fitting hose
    Diameter (Metric) Inner 1.27cm
    For Use With (Equipment) W4K Constant flow supplied air pump system
    Length (English) 100 ft.
    Length (Metric) 30.48m
    Diameter (English) Inner 0.5 in.
    Includes Schrader fittings

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit