Learn More
Selleck Chemical LLCSupplier Diversity Partner Spermidine trihydrochloride DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S365325MG
Spermidine, a natural polyamine produced from putrescine and decarboxylated S-adenosylmethionine (dcSAM) by spermidine synthase, is a novel autophagy inducer and negatively modulates N-methyl-d-aspartate (NMDA).
Catalog No. 50-194-8340