Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermidine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
100g
Packaging:
Glass bottle
124-20-9
C7H19N3
145.25
MFCD00008229
ATHGHQPFGPMSJY-UHFFFAOYSA-N
spermidine,1,5,10-triazadecane,4-azaoctamethylenediamine,spermidin,4-azaoctane-1,8-diamine,n1-3-aminopropyl butane-1,4-diamine,1,4-butanediamine, n-3-aminopropyl,n-3-aminopropyl butane-1,4-diamine,n-3-aminopropyl-1,4-butane-diamine,1,4-diaminobutane, n-3-aminopropyl
1102
CHEBI:16610
N'-(3-aminopropyl)butane-1,4-diamine
C(CCNCCCN)CN
124-20-9
C7H19N3
145.25
MFCD00008229
ATHGHQPFGPMSJY-UHFFFAOYSA-N
spermidine,1,5,10-triazadecane,4-azaoctamethylenediamine,spermidin,4-azaoctane-1,8-diamine,n1-3-aminopropyl butane-1,4-diamine,1,4-butanediamine, n-3-aminopropyl,n-3-aminopropyl butane-1,4-diamine,n-3-aminopropyl-1,4-butane-diamine,1,4-diaminobutane, n-3-aminopropyl
1102
CHEBI:16610
N'-(3-aminopropyl)butane-1,4-diamine
C(CCNCCCN)CN
Specific Gravity 0.925
Color Colorless
Boiling Point 128.0°C to 130.0°C (14.0mmHg)
Density 0.9250g/mL
Infrared Spectrum Authentic
Melting Point 22.0°C to 25.0°C
Refractive Index 1.4785 to 1.4805
Flash Point >110°C
Quantity 1g
Packaging Glass bottle
Assay Percent Range 99%
Beilstein 04,II,704
Merck Index 15,8869
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and ether
Formula Weight 145.25
Physical Form Liquid After Melting
Percent Purity 99%
Chemical Name or Material Spermidine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-689-