Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermidine trihydrochloride, 99+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
334-50-9
C7H19N3·3HCl
254.63
MFCD00012918
LCNBIHVSOPXFMR-UHFFFAOYSA-N
spermidine trihydrochloride,n-3-aminopropyl-1,4-butanediamine trihydrochloride,spermidine hydrochloride,n1-3-aminopropyl butane-1,4-diamine trihydrochloride,unii-1o14bed398,1,4-butanediamine, n-3-aminopropyl-, trihydrochloride,spermidine, trihydrochloride,1,4-butanediamine, n-3-aminopropyl , hydrochloride,n-3-aminopropyl butane-1,4-diamine trihydrochloride,4-azoniaoctamethylenediammonium trichloride
9539
N'-(3-aminopropyl)butane-1,4-diamine;trihydrochloride
C(CCNCCCN)CN.Cl.Cl.Cl
334-50-9
C7H19N3·3HCl
254.63
MFCD00012918
LCNBIHVSOPXFMR-UHFFFAOYSA-N
spermidine trihydrochloride,n-3-aminopropyl-1,4-butanediamine trihydrochloride,spermidine hydrochloride,n1-3-aminopropyl butane-1,4-diamine trihydrochloride,unii-1o14bed398,1,4-butanediamine, n-3-aminopropyl-, trihydrochloride,spermidine, trihydrochloride,1,4-butanediamine, n-3-aminopropyl , hydrochloride,n-3-aminopropyl butane-1,4-diamine trihydrochloride,4-azoniaoctamethylenediammonium trichloride
9539
N'-(3-aminopropyl)butane-1,4-diamine;trihydrochloride
C(CCNCCCN)CN.Cl.Cl.Cl
Melting Point 248°C to 252°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Assay Percent Range 99% min. (Argentometry)
Linear Formula H2N(CH2)4NH(CH2)3NH2·3HCl
Beilstein 04, II, 704
Merck Index 15, 8869
Solubility Information Solubility in water: soluble
Formula Weight 254.63
Physical Form Crystalline Powder
Percent Purity 99+%
Chemical Name or Material Spermidine trihydrochloride, 99+%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 206-379-0

RTECSNumber : EJ7023000

TSCA : TSCA