Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spermine NONOate, Tocris Bioscience™
Click to view available options
10mg
Description
A stable complex of nitric oxide and spermine used to generate a controlled release of nitric oxide in solution (EC50 = 6.2 μM for relaxation of rabbit aorta, and t½ =39 min at 37°C, pH 7.4, aqueous solution).
Specifications
CAS | 136587-13-8 |
Molecular Formula | C10H26N6O2 |
Quantity | 10 mg |
Synonym | spermine nonoate, spermine-nitric oxide, spermine nitric oxide complex, noc 22, noc-22, spermine nitric oxide adduct, spermine/nitric oxide complex, ccris 4091, sper/no, sper-no |
InChI Key | QPIOUFBAROSIAN-UHFFFAOYSA-N |
SMILES | C(CCN(CCCN)N(N=O)O)CNCCCN |
IUPAC Name | N-[3-aminopropyl-[4-(3-aminopropylamino)butyl]amino]-N-hydroxynitrous amide |
Molecular Weight (g/mol) | 262.358 |
PubChem CID | 5259 |
Formula Weight | 262.35 |
Show More |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction