Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPESP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549141

Catalog No. PA549141

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The encoded protein is a human alloantigen involved in sperm-egg binding and fusion. [provided by RefSeq, Apr 2010]
Specifications

Specifications

SPESP1
Polyclonal
PBS with 0.09% sodium azide
Q6UW49
Sperm equatorial segment protein 1, ESP, Equatorial segment protein, SP-ESP, Glycosylated 38 kDa sperm protein C-7/8, SPESP1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
246777
Western Blot
Unconjugated
SPESP1
Liquid
SPESP1
KLH conjugated synthetic peptide between 236-262 amino acids from the C-terminal region of human SPESP1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit