Learn More
SPEX CertiPrep™ Method AOAC 2007.01 SPEXQuE™ QuEChERS Kit
Sensible Solutions for Pesticide Extraction
Specifications
For Use With (Application) Sensible solutions for pesticide extraction
Methods AOAC 2007.01
View More Specs
Products 1
Catalog Number Mfr. No. Analyte or Component Names Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Analyte or Component Names Quantity Price Quantity & Availability  
AOACPSA2ML
View Documents Promotion Details
SPEX CertiPrepSupplier Diversity Partner
AOACPSA2ML
50mg PSA, 150mg MgSO4 2mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Meeting all your QuEChERS needs with general purpose kits suitable for removal of polar organic acids, some low to moderate levels of sugars and lipids.
  • US (AOAC Method 2007.01)
Specifications
Specifications
Sensible solutions for pesticide extraction
AOAC 2007.01