Learn More
SPEX™ SamplePrep 6801 Large Grinding Vial Set Accessory, 6801C4 Large Polycarbonate Center Cylinder
Replacement polycarbonate cylinders for 6801 Large Grinding Vial Set and 6871 Large low-chrome Grinding Vial Set
$539.00 - $2204.00
Specifications
Product Type Replacement Center Cylinder
For Use With (Equipment) 6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set
Material Polycarbonate
View More Specs
Products 2
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
6801C20
View Documents
SPEX™ SamplePrep
6801C20
20 Pk.
Pack of 20 for $2,204.00
Add to cart
6801C4
View Documents
SPEX™ SamplePrep
6801C4
4 Pk.
Pack of 4 for $539.00
Add to cart
Description
Description
  • Withstands liquid nitrogen temperatures
  • Transparent plastic allows visual checking of grinding progress
  • Disposable or may be cleaned with hot water and detergent or RNase/DNase-free solution
  • Sold as pack of 4 or 20
Specifications
Specifications
Replacement Center Cylinder
6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set
Polycarbonate