Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPEX™ SamplePrep 8000 Series Mixer/Mill™ Accessories Available on GSA/VA Contract for Federal Government customers only.

For use with 8000D and 8000M Mixer/Mills

Manufacturer:  SPEX™ SamplePrep 6133PC

 View more versions of this product

Catalog No. 6133PC


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Element C
1.9cm
Ideal for pulverizing soft, brittle materials and for mixing liquids and powders, slurry grinding
2 in.
Slip on Polyethylene cap
5mL
0.16 oz.
0.4 oz.
8000D and 8000M Mixer/Mills
0.75 in.
Yes
1.5
5.08cm
0.16 oz.
Polycarbonate
5mL
12mL
Grinding Vial
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit