Learn More
SPEX™ SamplePrep Prep-Aid™ Grinding Aids Available on GSA/VA Contract for Federal Government customers only.
Ideal for samples which crumble or ablate after pressing.
Manufacturer:  SPEX™ SamplePrep 3644150
 View more versions of this product
Catalog No. 3644-150
  Check Availability
Add to cart
Specifications
Description
  • Blends quickly and completely with samples at 10 to 25% by weight to form XRF sample disks
  • Moisture resistant and does not contaminate samples
  • Bonds 200 to 250 sample disks or provides matrix for 25 to 35 supported sample disks
Cellulose Binder
  • Can be used undiluted as a sample matrix with the sleeve-and-plunger technique
  • Does not cake in the bottle during storage
UltraBind™
  • Can be used undiluted as a sample matrix with the sleeve-and-plunger technique
  • Moisture resistant and resists cracking and swelling in storage
  • Includes low levels of NaCl
Paraffin Binder
  • Forms air-stable sample disks as the waxy binder seals the surface against moisture
Description & Specifications
Specifications
Samples which crumble or ablate after pressing
150g
Ultrabind
≤20μm Powder