Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPG21 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259135

Catalog No. NBP259135


Add to cart

Description

Description

SPG21 Polyclonal antibody specifically detects SPG21 in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

SPG21
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
Acid cluster protein 33, ACP33Spastic paraplegia 21 protein, GL010, MASPARDIN, MASTBM-019, spastic paraplegia 21 (autosomal recessive, Mast syndrome), Spastic paraplegia 21 autosomal recessive Mast syndrome protein
Rabbit
IgG
100 ul
Neurodegeneration
Primary
51324
Immunoprecipitation
Immunoprecipitation
Affinity Purified
SPG21
Maps to a region between 200 and 250
Immunogen affinity purified
RUO
Store at 4C.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only