Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPG7 Mouse anti-Human, Mouse, Clone: OTI1C12, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504420

Catalog No. 50-167-5114


Add to cart

Description

Description

Putative ATP-dependent zinc metalloprotease.
Specifications

Specifications

SPG7
Monoclonal
0.86 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q3ULF4, Q9UQ90
CAR, CMAR, PGN, SPG5C
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
234847, 6687
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1C12
Unconjugated
SPG7
Liquid
SPG7
Human recombit protein fragment corresponding to amino acids 300-573 of human SPG7 prodduced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit