Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPG7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548380

Catalog No. PA548380

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Putative ATP-dependent zinc metalloprotease.
Specifications

Specifications

SPG7
Polyclonal
PBS with 0.09% sodium azide
Q9UQ90
Paraplegin, 3424-, Spastic paraplegia 7 protein, SPG7, CAR, CMAR, PGN
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6687
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SPG7
Liquid
SPG7
KLH conjugated synthetic peptide between 115-141 amino acids from the Central region of human SPG7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit